Learn More
Electron Microscopy Sciences Reticle, Square Grid With 1 mm Divisions
SDP
Supplier:  Electron Microscopy Sciences A9100SQ
Encompass_Preferred
Reticles for use with our compound and inverted light microscopes. Reticle with square grid with 1 mm divisions for use with compound microscopes.
Catalog No. 50-342-70