Learn More
Apexbio Technology LLC Retigabine dihydrochloride, 5mg. Cas: 150812-13-8 MFCD: MFCD00912289. Antiepileptic compound
SDP
Supplier:  Apexbio Technology LLC A37585
Encompass_Preferred
Retigabine (D-23129) is a novel antiepileptic compound with broad spectrum and potent anticonvulsant properties, both in vitro and in vivo. The compound was shown to activate a K+ current in neuronal cells. 150812-13-8. MFCD00912289
Catalog No. 50-101-1820