Learn More
Selleck Chemical LLC Retinoic acid (Tretinoin) S1653-1g
SDP
Supplier:  Selleck Chemical LLC S16531G
Encompass_Preferred
Retinoic acid (Tretinoin) which is a ligand for both the retinoic acid receptor (RAR) and the retinoid X receptor (RXR) can induce granulocytic differentiation and apoptosis in acute promyelocytic leukemia (APL) cells Tretinoin (NSC 122758) can be used to induce animal models of osteoporosis
Catalog No. 50-310-8680