Learn More
Medchemexpress LLC Retinoic acid (all-trans retinoic acid) | 302-79-4 | MFCD00001551 | 99.7% | 300.44 g/mol | C20H28O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14649R5MG
Encompass_Preferred
Retinoic acid (all-trans-retinoic acid) analytical standard intended for research and analytical use. Provided as a high-purity reference material (CAS 302-79-4) for method development, calibration, and quantitative analysis by HPLC, GC, and MS. Suitable for assay validation and laboratory reference workflows.
  • High purity (99.73%) for accurate, reproducible results.
  • Supplied in small, convenient quantities for analytical testing.
  • Suitable for HPLC, GC, and mass spectrometry method development.
  • Useful for assay calibration, quality control, and validation.
  • Stable when handled and stored according to SDS instructions.
Catalog No. 50-002-23028