Learn More
American Radiolabeled Chemicals Inc RETINOL-11 12-3H 250UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 0566250UC
Encompass
Retinol-11,12-3H(N) 250uCi in ethanol under argon, 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 286.4, Shipped in Dry Ice, (Vitamin A alcohol)
Catalog No. 50-901-04541
Request a Quote
This product is regulated by the federal government, and may not be ordered online.