Learn More
Sigma Aldrich Fine Chemicals Biosciences Retinyl palmitate potency5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences R15125G
Encompass
Retinyl palmitate or vitamin A palmitate is a retinoid which is an acid form of vitamin A. Retinoids exist as many geometric isomers due to the unsaturated bonds in the aliphatic chain. Retinyl palmitate is a major storage form of vitamin A found in epidermis. Retinyl esters provide pools of vitamin A that are converted into retinol and other retinoids as needed.
Catalog No. 50-289-7126