Learn More
McMaster-Carr Cam Latches, Key Number 224
Supplier:  McMaster-Carr 1324A1
Encompass
The McMaster-Carr 224 key-number cam latch is a heavy duty latch made of steel with a zinc finish. It has a 1/4 turn cam action with a key-locking feature. The latch has a tongue width of 1-1/4", a tongue length of 2-1/2", and a mounting hole spacing of 2-3/4" x 1-1/8". It is rated for frequent access doors and panels on industrial equipment with a static load capacity of 175 lbs and a dynamic load capacity of 70 lbs. The cam latch comes with mounting screws and can be mounted on doors up to 1/2" thick.
Catalog No. NC2591944