Learn More
Med Vet International Von Klaus Gelpi Retractor, 7.5in, German Grade, Standard Pattern, Each
Supplier:  Med Vet International VK1244018
Encompass_Preferred
This Gelpi Retractor is a German grade surgical instrument. It features a single sharp tip on both ends and it is ratcheted. Often used in smaller surgical sites. A Gelpi retractor (self-retaining) is used to retract shallow incisions. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2665