Learn More
Medchemexpress LLC HY-114698 10mg Medchemexpress, Retro-2 cycl CAS:1429192-00-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11469810MG
Encompass_Preferred
Medchemexpress, HY-114698 10mg Retro-2 cycl CAS:1429192-00-6 Retro-2 cycl (RN 1-001) is a dihydroquinazolinone (DHQZ) inhibitor of retrograde trafficking. Retro-2 cycl (RN 1-001) inhibits JCPyV and HPV16 pseudovirus with IC50s of 54 μM and 160 μM, respectively. Antiviral agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9368