Learn More
Takara Bio RetroNectin® Recombinant Human Fibronectin Fragment, 2.5 mg
SDP
Supplier:  Takara Bio T100B
Encompass
RetroNectin reagent is a 63 kD fragment of recombinant human fibronectin fragment (also referred to as rFN-CH-296) that enhances the efficiency of lentiviral- and retroviral-mediated gene transduction. This is particularly important for hematopoietic cells and other hard-to-transfect cell types. Enhanced transduction is thought to result from co-localization of virus particles and target cells. This is accomplished by direct binding of viral particles to sequences in the heparin-binding domain and interaction of target-cell integrins with two other domains in rFN-CH-296. 2.5mg
Catalog No. 50-444-032