Learn More
Perkin Elmer US LLC Reusable Dessicant Kit - Qty. 4
Supplier:  Perkin Elmer US LLC L1250311
Encompass_Preferred
FT-IR spectrometers require dessicant kits to keep their optical paths moisture-free. PerkinElmer recommends that your instrument's desiccant be replaced every six months. Your spectrometer's manual has instructions on how to install the dessicant. Depending on the spectrometer, you will need two to three dessicant kits every six months.
Catalog No. 50-211-3582