Learn More
GLOMED INC Reusable Animal Feeding Needle 22G x 25mm Curved
Supplier:  GLOMED INC AFN2225C
Encompass_Preferred
Reusable Metal Hub with luer lock fitting - Smooth stainless steel ball tip and stainless steel tubing - Can be used as perfusion needle - Packed individually - Sold unsterile - please sterilize before use
Catalog No. NC1197789