Learn More
Medchemexpress LLC Reutericycline | 303957-69-9 | 98.1% | 349.46 | C20H31NO4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10324925MG
Encompass_Preferred
Reutericyclin is a tetramic-acid antibiotic produced by Lactobacillus reuteri and supplied as a research reagent for antimicrobial and biochemical studies. It has formula C20H31NO4 and a molecular weight of 349.46 g/mol. Supplied as a 25 mg solid, the compound is highly pure and soluble in DMSO for in vitro and in vivo formulation use.
  • Tetramic-acid antibiotic for antimicrobial research.
  • High purity suitable for biochemical and cellular assays.
  • Provided as a small solid quantity for bench-scale experiments.
  • Readily soluble in DMSO; ultrasonic agitation recommended.
  • Includes in vivo formulation protocols for suspension preparations.
  • Stable when stored as powder at -20°C for long-term storage.
Catalog No. 50-002-46648