Learn More
ABclonal Technology Rev-erb beta/NR1D2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A22050100UL
Encompass_Preferred
This gene encodes a member of the nuclear hormone receptor family specifically the NR1 subfamily of receptors The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism Alternatively spliced transcript variants have been described
Catalog No. 50-000-07512