Learn More
Selleck Chemical LLC Revaprazan Hydrochloride S5058-25mg
SDP
Supplier:  Selleck Chemical LLC S505825MG
Encompass_Preferred
Revaprazan Hydrochloride (YH1885) is a new reversible proton pump inhibitor with long-lasting acid-suppressive effects Revaprazan Hydrochloride reversibly inhibits H( )/K( )-ATPase via binding to the K -binding site of the pump
Catalog No. 50-311-1962