Learn More
NEOGEN Reveal for E. coli O157:H7, 20 Hour Complete System
SDP
Supplier:  NEOGEN 700002809
Encompass_Preferred
The Reveal for E. coli O157:H7 test system provides for the rapid recovery of Escherichia coli O157:H7 in foods, allowing detection and presumptive identification of the organism in as little as 8 hours for 25 g samples.Approvals: AOAC Official Method (including 375 g for raw beef product), No. 2000.13, No. 2000.14 (20-hour enrichment)Limit of Detection: 1 cfu/analytical unitTesting Time: 15 MinutesFor more information visit https://www.neogen.com/solutions/microbiology/reveal-e-coli-o157-h7/
Catalog No. 50-201-5128