Learn More
Abcam Reversan, MRP1 and P-glycoprotein inhibitor, 50MG
Supplier:  Abcam AB14586950MG
Encompass_Preferred
MW 441.53 g/mol, Purity >99%. Selective MRP1 and P-glycoprotein inhibitor. Chemosensitizing agent. Increases the sensitivity of MRP1-overexpressing tumor cells to doxorubicin (ab120629), vincristine (ab120226) and etoposide (ab120227). Shows chemosensitizing, antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5054