Learn More
Medchemexpress LLC Reverse T3-13C6 hydrochloride (3′,5′,3-Triiodothyronine-13C6 hydrochloride) | 1217676-14-6 | 96% | 693.39 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW010696AS1MG
Encompass_Preferred
Reverse T3-13C6 (hydrochloride) is the 13C labeled Reverse T3. It is used for research purposes and can serve as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-51905