Learn More
Medchemexpress LLC Reverse transcriptase-IN-1 | 2380001-43-2 | 98.1% | C25H17N7O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13024110MG
Encompass_Preferred
Reverse transcriptase-IN-1 is a diarylbenzopyrimidine analogue used as a potent, orally active HIV-1 nonnucleoside reverse transcriptase inhibitor for research applications. Supplied as a light yellow solid with high reported purity, it is intended for preclinical antiviral studies and biochemical assays. Store as powder at -20°C; solutions are stable at -80°C for six months or -20°C for one month.
  • Potent inhibition of HIV-1 reverse transcriptase.
  • Suitable for in vitro and preclinical antiviral studies.
  • High reported purity (~98.1%) for reliable assay performance.
  • Supplied as a solid suitable for formulation into solution stocks.
  • Defined storage recommendations to preserve stability.
Catalog No. 50-002-59022