Learn More
Research Products International Corp Reversible Dissecting Table
SDP
Supplier:  Research Products International Corp 246705
Encompass
  • Made of high-density polyethylene
  • Heavy duty and stain resistant
  • Drain groove to contain fluids
  • Dimensional scales in inches and centimeters
  • Will not dull fine surgical blades
  • Double sided
A new and unique approach makes this dissecting board more convenient than any other found on the market today. It is no longer necessary to buy different sizes as this board offers a large surface on one side and two smaller ones on the other side.
Made of heavy-duty stain resistant thick polyethylene, it will last for years to come without changing shape, bending or swelling. Will not dull fine surgical blades. In order to contain fluids, a drain groove is carved all around the edge of the DissecTable™.
Side 1:
  • Large cutting area including dimensional scales in inches and centimeters
  • 60 x 80 mm grid made of 48 x 10 mm squares
Catalog No. 50-222-9695