Learn More
TARGETMOL CHEMICALS INC Rezafungin 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T127321MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Rezafungin (SP-3025) is a broad-spectrum and long-lasting echinocandin with potent antifungal activity against Candida, Aspergillus and Pneumocystis. Purity 99.62%
Catalog No. 50-287-5937