Learn More
Medchemexpress LLC HY-108009A 5mg Medchemexpress, Rezafungin (acetate) CAS:1631754-41-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108009A5MG
Encompass_Preferred
Medchemexpress, HY-108009A 5mg Rezafungin (acetate) CAS:1631754-41-0 Rezafungin acetate (Biafungin acetate) is a next-generation, broad-spectrum, and long-lasting echinocandin. Rezafungin acetate shows potent antifungal activity against Candida spp., Aspergillus spp., and Pneumocystis spp.. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0727