Learn More
Medchemexpress LLC Rezetecán (SHR9265) | 99.6% | 1195.27 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1474081MG
Encompass_Preferred
Rezetecán (SHR9265) is a topoisomerase I inhibitor that can be used to synthesize Trastuzumab rezetecan, an antineoplastic agent. It also functions as a cytotoxic payload (ADC Cytotoxin) in antibody-drug conjugates (ADCs).
  • Topoisomerase I inhibitor
  • Used to synthesize Trastuzumab rezetecan, an antineoplastic agent
  • Functions as a cytotoxic payload (ADC Cytotoxin) in antibody-drug conjugates (ADCs)
Catalog No. 50-003-60443