Learn More
Medchemexpress LLC Rezivertinib | 1835667-12-3 | 486.57 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY109189500MG
Encompass_Preferred
Rezivertinib (BPI-7711) is an orally active, highly selective and irreversible third-generation EGFR tyrosine kinase inhibitor (TKI). It demonstrates high potency against common activation EGFR and resistance T790M mutations. Rezivertinib also exhibits excellent central nervous system (CNS) penetration and has antitumor activity.
  • Orally active
  • Highly selective
  • Irreversible third-generation EGFR tyrosine kinase inhibitor
  • Potent against common activation EGFR mutations
  • Potent against resistance T790M mutations
  • Excellent central nervous system (CNS) penetration
  • Antitumor activity
Catalog No. 50-003-45053