Learn More
Selleck Chemical LLC Rezivertinib E4675-1G
SDP
Supplier:  Selleck Chemical LLC E46751G
Encompass_Preferred
Rezivertinib(BPI-7711) is a potent third-generation inhibitor of EGFR tyrosine kinase targeting both EGFR-sensitizing mutations and the EGFR T790M mutation Additionally it has excellent CNS penetration and has antitumor activity
Catalog No. 50-000-39318