Learn More
Medchemexpress LLC Rezvilutamide | 1572045-62-5 | 99.8% | 479.47 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137448100MG
Encompass_Preferred
Rezvilutamide (SHR3680) is an orally active androgen receptor antagonist used for the study of prostate cancer. It inhibits the nuclear translocation, transcriptional activity, overexpression, and secretion of prostate-specific antigen of the androgen receptor, and induces CYP3A4 and CYP2B6.
  • Orally active androgen receptor antagonist
  • Used for the study of prostate cancer
  • Inhibits nuclear translocation of androgen receptor
  • Inhibits transcriptional activity of androgen receptor
  • Inhibits overexpression and secretion of prostate-specific antigen
  • Induces CYP3A4 and CYP2B6
Catalog No. 50-003-48997