Learn More
Medchemexpress LLC Rezvilutamide (SHR3680) | 1572045-62-5 | 99.8% | 479.47 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13744850MG
Encompass_Preferred
Rezvilutamide (SHR3680) is an orally active androgen receptor antagonist used for the study of prostate cancer. It inhibits the nuclear translocation, transcriptional activity, overexpression, and secretion of prostate-specific antigen of androgen receptor in prostate cancer cells, and exhibits an inducing effect on CYP3A4 and CYP2B6.
  • Inhibits nuclear translocation and transcriptional activity
  • Reduces overexpression and secretion of prostate-specific antigen in prostate cancer cells
  • Induces CYP3A4 and CYP2B6
Catalog No. 50-003-51593