Learn More
Glass Expansion RF Coil Gold for Varian Series 1 Radial
SDP
Supplier:  Glass Expansion 709001003G
Encompass_Preferred
RF Coil Gold for Agilent (Varian) Liberty Series 1 Radial ICP-OES. Compatible with Agilent (Varian) Liberty Series 1 Radial ICP-OES. The RF coil is susceptible to corrosion in the torch box, which eventually lowers the conductivity of the coil. The greater the corrosion, the larger amount of energy is needed to produce a plasma of the same power. Coils in a corroded condition, increases the stress applied to other components in the RF generating system, and can shorten the lifetime of your torch. Copper is used as the base material of most commercially available coils but different plating materials are available. Silver has the best conductance & gold the best corrosion resistance. Only Gold is available for this ICP.To install your coil and ensure proper alignment, we highly recommend our re-useable RF Coil Installation Tool Kit (501907318).Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7316