Learn More
Glass Expansion RF Coil Installation Tool for Varian Series 1 Radial Suitable for: Liberty Series 1
SDP
Supplier:  Glass Expansion 709001003T
Encompass_Preferred
RF Coil Installation Tool for Agilent (Varian) Liberty Series 1 Radial ICP-OES. This tool is for use with all Glass Expansion’s replacement RF Coils for Agilent (Varian) Liberty Series 1 Radial ICP-OES, P/N 501907316. To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7318