Learn More
Glass Expansion RF Coil Installation Tool for Agilent 7500/7700/7800/7900/8800/8900
SDP
Supplier:  Glass Expansion 709007500T
Encompass_Preferred
RF Coil Alignment Tool for Agilent 7500/7700/7800/7900/8800/8900 ICP-MS. This tool is for use with the following Glass Expansion’s replacement RF Coils, P/Ns Copper 501907347, Gold 501907348, Silver 501907349. To install your RF coil and ensure proper alignment, we highly recommend using our re-useable RF Coil Installation Tool Kit.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7346