Learn More
MOLECULAR DEPOT LLC Alkali-soluble Casein - 50 mL
Supplier:  MOLECULAR DEPOT LLC B2012712
Encompass_Preferred
Alkali-soluble Casein (5perc) consists of a concentrated solution of alkali-soluble casein that is used as a unique blocking reagent with ultra-low background This product has been used as molecular tool for various biochemical applications It has also been used in a wide array of other chemical and immunological applications For in vitro research use only
Catalog No. 50-000-13414