Learn More
Medchemexpress LLC RF9 hydrochloride 50mg | 50MG
SDP
Supplier:  Medchemexpress LLC HY107382A50MG
Encompass_Preferred
RF9 hydrochloride is a potent and selective Neuropeptide FF receptor antagonist with Ki values of 58 and 75 nM for hNPFF1R and hNPFF2R respectively[1 [2
Catalog No. 50-002-39928