Learn More
Polyscience 15L Refrigerated Circulator, Advanced Digital Controller (-40° to 200°C); 120V, 50/60Hz
SDP
Supplier:  Polyscience AD15R40A11B
Encompass_Preferred
PolyScience™ 15 Liter Refrigerated CirculatorAdvanced Digital Controller-40 to 200° C Working Temperature RangeTemperature Stability: ±0.01°Cooling Capacity at 20° C: 1000W1100W HeaterMaximum Pressure: 4.3 psi (0.30 bar) Maximum Pressure Flow Rate: 5.3 gpm (20.1 LPM) Maximum Suction Flow Rate: 3.9 gpm (14.7 LPM)120V, 60Hz• Intuitive 3.75” (9.5 cm) display with touch-pad control• 4 languages: French, German, Spanish, English• Swivel 180™ Rotating Controller• On-board connectivity: USB-A & B, Ethernet, RS-232/RS-485, and external temperature probe• On-screen prompts• Automatic performance optimization and specific heat tuning• Single-point calibration capability
Catalog No. 50-113-0576
Only null left
Add to Cart
Request a Quote