Learn More
Polyscience 7L Space-Saving Refrigerated Circulator, Advanced Programmable Controller (-20 to 200°C); 120V, 60Hz
SDP
Supplier:  Polyscience AP07R20A11B
Encompass_Preferred
PolyScience™ 7 Liter Space-Saving Refrigerated CirculatorAdvanced Programmable Controller-20 to 200°C Working Temperature RangeTemperature Stability: ±0.005°CCooling Capacity at 20°C: 200W1100W HeaterMaximum Pressure: 4.3 psi (0.30 bar) Maximum Pressure FlowRate: 5.3 gpm (20.1 LPM) 120V, 60HzFeatures:• OpenMode time time/temperature programming (no restrictions on number of programs or steps)• Intuitive 4.3” (10.9 cm) SmartTouch display• 11 languages: French, German, Spanish, Chinese, Portuguese, Russian, Hindi, Arabic, Italian, Korean, English• Variable speed pump with open- and closed-loop external circulation capability• Swivel 180™ Rotating Controller• On-board connectivity: USB-A & B, Ethernet, RS-232/RS-485, remote on/off and external temperature probe• Event scheduling (time & date) with real-time clock• Review temperature trends for up to 10 days• Multiple selectable “home” screens• On-screen help
Catalog No. 50-113-0544
Add to Cart
Request a Quote