Learn More
Cell Biolabs Inc RFP ELISA Kti, 96 assays
Supplier:  Cell Biolabs Inc AKR122
Encompass_Preferred
Although RFP expression can be detected under a fluorescence microscope, RFP fluorescence intensity varies from cell to cell because of the heterogeneity nature of RFP expression. In order to quantitate the RFP expression in cells, FACS analysis is usually required, which is both expensive and time consuming. Cell Biolabs’ RFP ELISA Kit is an enzyme immunoassay developed for quantitation of RFP or RFP fusion protein in cell or tissue samples. The quantity of RFP or its variants (including TagRFP, TurboRFP, DsRed, tdTomato, mCherry, mKate, mRuby, mBanana, mOrange, mPlum, and mStrawberry) in an unknown sample is determined by comparing its absorbance with that of a known RFP standard curve. The kit has detection sensitivity limit of 150 pg/mL RFP. The kit also provides an efficient system for rapid quantitation of RFP lentivirus titer for both viral supernatant and purified virus. Each kit provides sufficient reagents for up to 96 assays including standard curve and RFP samples.
Catalog No. 50-672-578