Learn More
Medchemexpress LLC HY-101676 5mg Medchemexpress, RG-12525 CAS:120128-20-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1016765MG
Encompass_Preferred
Medchemexpress, HY-101676 5mg RG-12525 CAS:120128-20-3 RG-12525 is a a specific, competitive and orally effective antagonist of the peptidoleukotrienes, LTC4, LTD4 and LTE4, inhibiting LTC4-, LTD4- and LTE4-inducd guinea pig parenchymal strips contractions, with IC50s of 2.6 nM, 2.5 nM and 7 nM, respectively; RG-12525 is also a peroxisome proliferator-activated receptor gamma (PPAR-gamma) agonist with IC50 of appr 60 nM and a potent inhibitor of CYP3A4, with a Ki value of 0.5 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0641