Learn More
Selleck Chemical LLC RG-7112
SDP
Supplier:  Selleck Chemical LLC S703010MM/1ML
Encompass_Preferred
RG7112 (RO5045337) is an orally bioavailable and selective p53-MDM2 inhibitor with HTRF IC50 of 18 nM
Catalog No. 50-000-02752