Learn More
OTT Hydromet Adcon - RG1-400
Supplier:  OTT Hydromet 200.733.048
Encompass_Preferred
This version of the RG1 weighs in at approx. 750gr. and is suited for both mast and floor mounting. With its large orifice of 400cm² and its high resolution of 0.1mm it is ideally suited for areas with light to moderate rainfall intensity. The measurement is done by a double tipping bucket system that's factory calibrated to remain within a <2% accuracy in rain falls up to 30mm per hour. The steep angle of the buckets makes water run of quickly and thus prevents the build-up of residue, like sand or dust in the buckets. An aluminum filter can easily be removed for cleaning. The funnel attaches to the body with a bayonet-style lock, which prevents the funnel from being shaken off in heavy winds. The rain gauges exterior is fully made of aluminum that's anodized for extra protection and durability. The sensor comes complete with all mast mounting equipment, two pipe clamps, and a shielded 1m cable with a 7-pin Binder jack, compatible to all RTU's of the A723, A733 and A75x families.
Catalog No. NC1919661
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.