Learn More
Active Motif RG108
SDP
Supplier:  Active Motif 14105
Encompass
Active Motif's RG108 is a non-nucleoside DNA methyltransferase inhibitor (IC50 = 115 nM). It inhibits DNA methylation in human and tumor cell lines without toxicity, and enhances the reversion of neural progenitor cells to the pluripotent state.
Catalog No. NC1841008