Learn More
Cayman Chemical RG33trIfluoroacetate salt 10mg
SDP
Supplier:  Cayman Chemical 3751610 MG
Encompass_Preferred
An ApoA1 peptidomimetic; solubilizes DMPC-containing MLVs to form recombinant HDL; lipid-bound RG33 induces cholesterol efflux in J774 macrophages; decreases blood glucose levels in insulin-resistant mice at 12 mg/kg
Catalog No. 50-291-6141