Learn More
Apexbio Technology LLC RG7112, 10mg. Cas: 939981-39-2 MFCD: MFCD18206914. MDM2 inhibitor, first clinical
SDP
Supplier:  Apexbio Technology LLC A376210
Encompass_Preferred
RG7112 is a selective inhibitor of p53-MDM2 binding that frees p53 from negative control, activating the p53 pathway in cancer cells leading to cell cycle arrest and apoptosis. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4837