Learn More
Medchemexpress LLC RG7697 (NNC0090-2746) | 99.9% | 4473.04 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1027110MG
Encompass_Preferred
RG7697 is a dual agonist for glucagon-like peptide receptor (GLP Receptor) and glucose-dependent insulinotropic polypeptide receptor (GIPR), with EC50 values of 5 pM and 3 pM, respectively. This compound exhibits antihyperglycemic properties and is intended for research use.
  • Increases insulin secretion and metabolism
  • Decreases glucose excursion
  • Ameliorates diet-induced type 2 diabetes and obesity in C57BL/6 mice
  • Reduces body weight, blood glucose, and food intake
  • Decreases intraperitoneal glucose tolerance, plasma insulin, and cholesterol
  • For research use only
Catalog No. 50-004-25138