Learn More
Aviva Systems Biology Corp RG9MTD2 Antibody - middle region (ARP41144_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41144_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RG9MTD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RG9MTD2. Alternate names: TRM10, RG9MTD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57736