Learn More
Medchemexpress LLC RGD peptide (GRGDNP) | 114681-65-1 | 99.83% | 614.61 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP174050MG
Encompass_Preferred
RGD peptide (GRGDNP) is an inhibitor of integrin-ligand interactions. It competitively inhibits α5β1 binding with extracellular matrix (ECM) and promotes apoptosis through the activation of conformational changes that enhance pro-caspase-3 activation and autoprocessing. This peptide plays an important role in cell adhesion, migration, growth, and differentiation.
  • Inhibits integrin-ligand interactions.
  • Competitively inhibits α5β1 binding with extracellular matrix (ECM).
  • Promotes apoptosis through activation of conformation changes that enhance pro-caspase-3 activation and autoprocessing.
  • Important for cell adhesion, migration, growth, and differentiation.
Catalog No. 50-004-23151