Learn More
CPC Scientific H-Arg-Gly-Asp-Ser-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific RGDP010A
Encompass
RGDS is a tetrapeptide that supports fibroblast attachment and inhibition of fibronectin binding to platelets.SEQUENCE: H-Arg-Gly-Asp-Ser-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: RGDSMOLECULAR FORMULA: C15H27N7O8MOLECULAR WEIGHT: 433.42STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [91037-65-9]SYNONYMS: RESEARCH AREA: CancerREFERENCES:SKU(s): RGDP-010A
Catalog No. 50-211-0385