Learn More
Apexbio Technology LLC RGFP966, 1357389-11-7, MFCD26960954, 25 mg
SDP
Supplier:  Apexbio Technology LLC A880325
Encompass_Preferred
MF: C21H19FN4O, MW: 362.4, Solubility: 18.12mg/mL in DMSO. RGFP966 is a selective inhibitor of HDAC3 with IC50 value of 80 nM.
Catalog No. 50-101-5192