Learn More
Aviva Systems Biology Corp Rgs1 Antibody - N-terminal region (ARP42843_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42843_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Rgs1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: BL34ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-034