Learn More
Hach Company Phosphorus Reagent Set, Ascorbic Acid Rapid Liquid™
Supplier:  Hach Company 2678600
Encompass
For determination of reactive Phosphorus (ortho-phosphate) by the Ascorbic Acid Rapid Liquid™ method. Hach Method 10055. Range: 19 - 3000 μg/L PO4. Set includes: Ascorbic Acid Reagent Dilution Solution, Ascorbic Acid Reagent Powder and Molybdate Reagent Solution for approximately 250 tests.
Catalog No. 50-199-6536