Learn More
Hach Company Nitrogen-Ammonia Reagent Set, TNT, AmVer (Salicylate),Low Range
Supplier:  Hach Company 2604545
Encompass
For determination of Low Range Ammonia Nitrogen by the AmVer™ Salicylate Test 'N Tube™ Method. Hach Method 10023. Range: 0.02 - 2.50 mg/L NH3-N. Reagent set contains: AmVer Diluent Reagent Low Range Test 'N Tubes, Ammonia Salicylate. Reagent Powder Pillows and Ammonia Cyanurate Reagent Powder Pillows. 25-50 tests (dependent upon the number of reagent blank tests performed)
Catalog No. 50-199-6325