Learn More
Hach Company NitraVer X Nitrogen-Nitrate Reagent Set, HR
Supplier:  Hach Company 2605345
Encompass
For determination of High Range Nitrate Nitrogen by the Chromotropic Acid Test 'N Tube Method. Hach Method 10020. Range: 0.2 - 30.0 mg/L NO3-N 50 tests per package.
Catalog No. 50-199-6328